missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Yeast Lytic Enzyme, Arthrobacter luteus
Description
Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Off-white |
| Product Type | Cell Lysis Enzyme |
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| For Use With (Application) | Hydrolyzes linear glucose polymers with beta-1,3-linkages |
| Quantity | 250 mg |
Sécurité et manipulation
| Recommended Storage | Keep cold |
RUO – Research Use Only