missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast Lytic Enzyme, Arthrobacter luteus

Catalog No. AAJ63195MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63195MD 250 mg
AAJ63195ME 500 mg
AAJ6319503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63195MD Supplier Thermo Scientific Chemicals Supplier No. J63195MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrolyzes linear glucose polymers with beta-1,3-linkages | CAS: 37340-57-1

Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37340-57-1
Synonym Lyticase; Zymolyase 20T

Specifications

Color Off-white
Product Type Cell Lysis Enzyme
Content And Storage 2°C to 8°C
Form Lyophilized
For Use With (Application) Hydrolyzes linear glucose polymers with beta-1,3-linkages
Quantity 250 mg

RUO – Research Use Only