missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ YE 120
Description
GPR35 agonist (EC50 = 32 nM in DMR assays). Also displays partial agonist activity in a β-arrestin translocation assay (EC50 = 10.2 μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 330.17 |
| Purity | >98% |
| Molecular Weight (g/mol) | 330.168 |
| Chemical Name or Material | YE 120 |
Safety and Handling
| Recommended Storage | Store at -20°C |