accessibility menu, dialog, popup

UserName

CH3NNOH3CClClN

CAS RN 383124-82-1

IUPAC Name: 2-[3-cyano-5-(3,4-dichlorophenyl)-4,5-dimethyl-2,5-dihydrofuran-2-ylidene]propanedinitrile
Synonyms: 2-[3-cyano-5-(3,4-dichlorophenyl)-4,5-dimethylfuran-2-ylidene]propanedinitrile
Molecular Weight (g/mol): 330.17
Molecular Formula: C16H9Cl2N3O
InChi Key: RSWZFAPYKOARNG-UHFFFAOYNA-N
SMILES: CC1=C(C#N)C(OC1(C)C1=CC(Cl)=C(Cl)C=C1)=C(C#N)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results