missing translation for 'onlineSavingsMsg'
Learn More

Xylose, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18607793
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607793 100 g Amber Glass Bottle
18607794 500 g Amber Glass Bottle
18607795 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607793 Supplier Spectrum Chemical Mfg Cor Supplier No. XY110100GM
Only null left

XY110, 58-86-6, C5H10O5

Spectrum™ Chemical Xylose, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-UHFFFAOYNA-N
IUPAC Name oxane-2,3,4,5-tetrol
SMILES OC1COC(O)C(O)C1O

Specifications

CAS Max % 1
Ignition Residue 0.0005
Loss on Drying 0.001
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +18.2° to +19.4°
Percent Purity 98 to 102%
Grade USP