missing translation for 'onlineSavingsMsg'
Learn More

Xylitol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015737
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015737 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015737 Supplier Merck Emd Millipore Supplier No. PHR12801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Xylitol is a naturally occurring polyhydric sugar alcohol with high sweetening power and anticariogenic properties.

Specifications

CAS 87-99-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOCH2 [CH(OH)]3 CH2 OH
MDL Number MFCD00064292
Melting Point 94°C to 97°C
Synonym Xylite
Type Certified Reference Material
Quantity 1 g
Formula Weight 152.15
Grade Certified Reference Material
Show More Show Less