missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, NF, 98.5-101%, Spectrum™ Chemical

Catalog No. 18607783
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607783 50 kg Poly Bottle
18607779 100 g Poly Bottle
18607780 500 g Poly Bottle
18607781 2.5 kg Poly Bottle
18607782 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18607783 Supplier Spectrum Chemical Mfg Cor Supplier No. X101750KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

X1017, 87-99-0, C5H12O5

Spectrum™ Chemical Xylitol, NF is used as a sugar substitute in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
IUPAC Name pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

CAS Max % 1
Boiling Point 216°C (420.8 °F)
Ignition Residue 0.005
Packaging Poly Bottle
Quantity 50 kg
Percent Purity 98.5 to 101%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Xylitol