missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylitol, FCC, 98.5-101%, Spectrum™ Chemical
X1064, 87-99-0, C5H12O5
$288.24 - $9472.58
Chemical Identifiers
| CAS | 87-99-0 |
|---|---|
| Molecular Formula | C5H12O5 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00064291,MFCD00064292 |
| InChI Key | HEBKCHPVOIAQTA-UHFFFAOYNA-N |
| IUPAC Name | pentane-1,2,3,4,5-pentol |
| SMILES | OCC(O)C(O)C(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18607774
|
Spectrum Chemical Mfg Cor
X1064100GM |
100 g | Poly Bottle |
Each for $288.24
|
|
||||
|
18607775
|
Spectrum Chemical Mfg Cor
X1064500GM |
500 g | Poly Bottle |
Each for $812.45
|
|
||||
|
18607776
|
Spectrum Chemical Mfg Cor
X106425KG |
2.5 kg | Poly Pail |
Each for $2,200.61
|
|
||||
|
18607777
|
Spectrum Chemical Mfg Cor
X106412KG |
12 kg | Poly Pail |
Each for $4,508.39
|
|
||||
|
18607778
|
Spectrum Chemical Mfg Cor
X106425KGBL |
25 kg | Fiber Drum |
Each for $9,472.58
|
|
||||
Description
Spectrum™ Chemical Xylitol, FCC is used as nutritive sweetener as a sugar substitute to produce âsugar-freeâ foods and is commonly used in the production of chewing gums, mints and other candies. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 87-99-0 | |
| 152.15 | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| OCC(O)C(O)C(O)CO |
| C5H12O5 | |
| MFCD00064291,MFCD00064292 | |
| pentane-1,2,3,4,5-pentol |
Specifications
| 87-99-0 | |
| 0.001 | |
| C5H12O5 | |
| 100 g | |
| OCC(O)C(O)C(O)CO | |
| 152.15 | |
| FCC |
| 1 | |
| Poly Bottle | |
| MFCD00064291,MFCD00064292 | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| pentane-1,2,3,4,5-pentol | |
| 98.5 to 101% |