missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, FCC, 98.5-101%, Spectrum™ Chemical

X1064, 87-99-0, C5H12O5

Supplier:  Spectrum Chemical Mfg Cor X106412KG

 View more versions of this product

Catalog No. 18607777


Only null left

Description

Description

Spectrum™ Chemical Xylitol, FCC is used as nutritive sweetener as a sugar substitute to produce “sugar-free” foods and is commonly used in the production of chewing gums, mints and other candies. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

87-99-0
0.001
C5H12O5
12 kg
OCC(O)C(O)C(O)CO
152.15
FCC
1
Poly Pail
MFCD00064291,MFCD00064292
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
98.5 to 101%

Chemical Identifiers

87-99-0
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
OCC(O)C(O)C(O)CO
C5H12O5
MFCD00064291,MFCD00064292
pentane-1,2,3,4,5-pentol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions