missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, FCC, 98.5-101%, Spectrum™ Chemical

Catalog No. 18607774
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607774 100 g Poly Bottle
18607775 500 g Poly Bottle
18607776 2.5 kg Poly Bottle
18607777 12 kg Poly Bottle
18607778 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18607774 Supplier Spectrum Chemical Mfg Cor Supplier No. X1064100GM

X1064, 87-99-0, C5H12O5

Spectrum™ Chemical Xylitol, FCC is used as nutritive sweetener as a sugar substitute to produce sugar-free foods and is commonly used in the production of chewing gums, mints and other candies. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
IUPAC Name pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

CAS Max % 1
Boiling Point 216°C (420.8°F)
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98.5 to 101%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Xylitol