missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenol Orange Indicator, 0.2% (w/v) in Alcohol, Ricca Chemical
Spécifications
Spécifications
| Nom ou substance chimique | Xylenol Orange |
| Concentration ou composition (par analyte ou composants) | 0.2% (w/v) Alcoholic Solution, in SDA-3A (100 parts Ethanol and 5 parts Methanol) |
| Emballage | Natural Poly Bottle |
| Quantité | 120 mL |
Sécurité et manipulation
| ShelfLife | 30 days |