missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange, 0.1% Aqueous, Certified, LabChem™

$374.93 - $472.00

Chemical Identifiers

CAS 3618-43-7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC270007
View Documents
LabChem
LC270007
125 mL Poly Bottle
Each for $374.93
Only null left
 
LC270001
View Documents
LabChem
LC270001
500 mL Poly Bottle
Each for $472.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3618-43-7

Specifications

3618-43-7 , 7732-18-5
C31H28N2Na4O13S
Passes Test
Certified
1g/mL
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Purple/Red
Liquid
99.9,0.1
Soluble in water
760.59
Poly Bottle
Passes Test
1g/mL
125 mL
Xylenol Orange, 0.1% Aqueous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents