missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange, 0.1% Aqueous, Certified, LabChem™

Supplier:  LabChem LC270001

 View more versions of this product

Catalog No. LC270001


Only null left

Description

Description

Specifications

Specifications

Xylenol Orange
3618-43-7 , 7732-18-5
C31H28N2Na4O13S
Passes Test
Certified
1g/mL
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Purple/Red
Liquid
0.1% Aqueous
99.9,0.1
Soluble in water
760.59
Poly Bottle
Passes Test
1g/mL
500 mL

Chemical Identifiers

3618-43-7
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions