missing translation for 'onlineSavingsMsg'
Learn More

Xylenes/Ethanol, 50/50 v/v, Reagents

Catalog No. 01333379
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333379 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333379 Supplier Labchem Inc Supplier No. CX1950004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Xylenes/Ethanol, 50/50 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1330-20-7 , 64-17-5 , 67-63-0
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.835
Grade Reagent
Chemical Name or Material Xylenes/Ethanol, 50/50 v/v