missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, contains ethylbenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Description
- For Microelectronic Use
Specifications
Specifications
| Quantity | 500 mL |
| Flash Point | 29.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 0.86kg |
| Molecular Weight (g/mol) | 106.17 |
| Formula Weight | 106.17 |
| Percent Purity | 98.5% (GC) |
| Grade | ACS |
| Chemical Name or Material | Xylenes, Contains ethylbenzene |