missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich Xylene Substitute

Catalog No. 01288041
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288041 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288041 Supplier Sigma-Aldrich Supplier No. A55971GAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with microscopy.

Compatible with all tissue processors. May not be compatible with all permanent mounting media. Xylene Substitute has been used to dewax or de-paraffinize the specimen sections after the staining process.

  • Sigma-Aldrich is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry
  • This product has Inherently Safer Chemistry, compared to the standard use of formalin for tissue processing.
  • Form: solution
  • Applications: hematology and histology
  • Storage temperature: 2°C to 8°C.

Specifications

Physical Form Solution
Color Clear
Quantity 1 gal.
Grade Reagent