missing translation for 'onlineSavingsMsg'
Learn More

Xylene Cyanole Indicator, 0.005%, Reagents

Catalog No. 01333380
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333380 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333380 Supplier Labchem Inc Supplier No. CX000500500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Xylene Cyanole Indicator, 0.005%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Xylene Cyanole Indicator, 0.005%
CAS 7732-18-5
Color Blue to Blue/Green
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1
Boiling Point 100°C
Percent Purity 0.005%
Grade Reagent