missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Xplorer™ Single-Channel Variable Pipette
GREENER_CHOICE

Catalog No. 05414049
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 mL
1 to 20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05414049 1 to 20 μL
05414050 0.1 to 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05414049 Supplier Eppendorf™ Supplier No. 4861000017

From blue sky research to clinical applications, you’re working on challenging problems every day. You need the best tools and the best laboratory equipment, so when pipetting, the Eppendorf Xplorer or Xplorer plus electronic pipettes offer simplicity, precision, and reproducibility.

  • Electronic air-cushion pipette is simple, precise and reproducible
  • Intuitive handling with multi-functional rocker and selection dial for quick and easy selection of functions
  • Self-explanatory color display without submenus and with ergonomic angle for optimal readability
  • Spring-loaded tip cone to minimize tip attachment forces to help to reduce stress
  • Rechargeable battery for up to 8 hours operation before recharge
  • Separate power socket so work can continue while charging
  • Easy adjustment to potentially variable conditions (e.g. geographic height, liquids with high density)
  • Lower part of pipette can be autoclaved to ensure decontamination
  • 9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish

Specifications

Battery Type Lithium Polymer
Certifications/Compliance Lot-specific Certificate
Compatible Tips Universal Tips, epT.I.P.S.™
Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With Pipetting of Aqueous Liquids
Model Single Channel Electronic
Number of Channels 1
Product Line Xplorer
Type Electronic Pipette
Volume (Metric) 1 to 20 μL
For Use With (Application) Pipetting of Aqueous Liquids
Increments 0.02 μL
Plug Type US
Amperage 1 A
Show More Show Less