missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xenon difluoride, 99.5+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Melting Point | 129°C |
| Color | White |
| Physical Form | Crystals |
| Quantity | 5 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 99.5+% |
| Linear Formula | XeF2 |
| Merck Index | 15, 10270 |
| Solubility Information | Solubility in water: decomposes in water |
| Formula Weight | 169.29 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. May cause fire or explosion; strong oxidizer. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep/Store away from clothing/ combustible materials. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 237-251-2 |
RUO – Research Use Only