missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XBP1 Recombinant Rabbit Monoclonal Antibody (23GB4840)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552749
Description
XBP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
XBP1 encodes a transcription factor that regulates MHC class II genes by binding to a promoter element referred to as an X box. This gene product is a bZIP protein, which was also identified as a cellular transcription factor that binds to an enhancer in the promoter of the T cell leukemia virus type 1 promoter. It may increase expression of viral proteins by acting as the DNA binding partner of a viral transactivator. It has been found that upon accumulation of unfolded proteins in the endoplasmic reticulum (ER), the mRNA of this gene is processed to an active form by an unconventional splicing mechanism that is mediated by the endonuclease inositol-requiring enzyme 1 (IRE1). The resulting loss of 26 nt from the spliced mRNA causes a frame-shift and an isoform XBP1(S), which is the functionally active transcription factor. The isoform encoded by the unspliced mRNA, XBP1(U), is constitutively expressed, and thought to function as a negative feedback regulator of XBP1(S), which shuts off transcription of target genes during the recovery phase of ER stress. A pseudogene of XBP1 has been identified and localized to chromosome 5.Identifiants chimiques
| XBP1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Xbp1 | |
| D11Ertd39e; Hepatocarcinogenesis-related transcription factor; HTF; OTTHUMP00000028682; Tax-responsive element-binding protein 5; tax-responsive element-binding protein 5 homolog; TREB5; TREB-5; X-box binding protein 1; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP1; XBP-1; XBP2 | |
| Xbp1 | |
| Primary | |
| Affinity chromatography |
| 23GB4840 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17861 | |
| 7494 | |
| A synthesized peptide derived from human XBP1 (120-160AA). | |
| Antibody |
Spécifications
| XBP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Xbp1 | |
| D11Ertd39e; Hepatocarcinogenesis-related transcription factor; HTF; OTTHUMP00000028682; Tax-responsive element-binding protein 5; tax-responsive element-binding protein 5 homolog; TREB5; TREB-5; X-box binding protein 1; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP1; XBP-1; XBP2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7494 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4840 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17861 | |
| Xbp1 | |
| A synthesized peptide derived from human XBP1 (120-160AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |