missing translation for 'onlineSavingsMsg'
Learn More

Xanthydrol, 98+%

Catalog No. AAA1347622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1347622 100 g
AAA1347606 5 g
AAA1347614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1347622 Supplier Thermo Scientific Chemicals Supplier No. A1347622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-46-0 | C13H10O2 | 198.221 g/mol

Useful in tests for urea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-46-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00005057
InChI Key JFRMYMMIJXLMBB-UHFFFAOYSA-N
Synonym 9-hydroxyxanthene, xanthydrol, xanthanol, 9-xanthydrol, xanthen-9-ol, xanthene, hydroxy, 9-xanthenol, ccris 1640, unii-7131m69ikf, xanthrol
PubChem CID 72861
IUPAC Name 9H-xanthen-9-ol
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)O

Specifications

Melting Point 122°C to 126°C
Quantity 100 g
Beilstein 10395
Sensitivity Light sensitive
Formula Weight 198.22
Percent Purity ≥98%
Chemical Name or Material Xanthydrol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-996-1
RTECSNumber ZD5710000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only