missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WWP1 Monoclonal Antibody (K24023_16C10), Invitrogen™

Numéro de catalogue. MA555811
Modifier l'affichage
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA555811 50 μL
MA556191 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA555811 Fournisseur Invitrogen™ Code fournisseur MA555811
Il en reste null

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KLERVKEQLK LSLENKNGIA QTGELTVVLD GLVIEQENIT NCSSSPTIEI QENGDALHEN GEPSARTTAR LAVEGTNGID NHVPTSTLVQ NSCCSYVVNG DNTPSSPSQV AARPKNTPAP KPLASEPADD TVNGESSSFA PTDNASVTGT

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 4 tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. The encoded protein belongs to a family of NEDD4-like proteins, which are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. Alternative splicing of this gene generates at least 6 transcript variants; however, the full length nature of these transcripts has not been defined. [provided by RefSeq].

Spécifications

Antigen WWP1
Applications Western Blot
Classification Monoclonal
Clone K24023_16C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Wwp1
Gene Accession No. Q9H0M0
Gene Alias 8030445B08Rik; AIP5; atrophin-1 interacting protein 5; atrophin-1-interacting protein 5; DKFZp434D2111; HECT-type E3 ubiquitin transferase WWP1; hSDRP1; NEDD4-like E3 ubiquitin-protein ligase WWP1; Nedd-4-like ubiquitin-protein ligase; SDRP1; TGIF-interacting ubiquitin ligase 1; Tiul1; Unknown (protein for MGC:129005); WW domain containing E3 ubiquitin protein ligase 1; WW domain-containing protein 1; Wwp1
Gene Symbols Wwp1
Host Species Mouse
Immunogen Recombinant human WWP1 (immunogen range is 111-260).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11059
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats