missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GHS Labels app for Brady Workstation

Catalog No. 19112937
Change view
Click to view available options
:
GHS Label App
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19112937 GHS Label App
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19112937 Supplier Brady™ Supplier No. BWRKGHSDWN
Only null left

For creating any size GHS label and print it any time. Brady™ GHS Labels app for Brady Workstation, also used for Secondary container labeling, Chemical labeling.

Get started quickly with a download: Receive the license key via email

  • Helps creating GHS labels of any size and print it at any time

Specifications

Product Type Software
For Use With (Equipment) BBP31 Sign and Label Printer, BBP33 Label Printer, BBP85 Industrial Sign and Label Printer, GlobalMark Label Maker
Languages English
For Use With (Application) Secondary container labeling, Chemical labeling