missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WM 3835
Description
WM 3835 is a lysine acetyltransferase HBO1 (KAT7) inhibitor. Reduces H3K14ac levels and inhibits growth and viability of acyte myeloid leukemia cell lines in vitro. Cell-permeable.
Spécifications
Spécifications
| Chemical Name or Material | N'-(4-Fluoro-5-methyl-[1,1 '-biphenyl]-3-carbonyl)-3-hydroxybenzenesulfonohydrazide |
| CAS | 2229025-70-9 |
| Quantity | 50 mg |
| Target | Histone Acetyltransferase Inhibitors |
| Molecular Formula | C20H17FN2O4S |
| Purity | 0.98 |