missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Wiskostatin
Description
Inhibitor of neural Wiskott-Aldrich syndrome protein (N-WASP) activity. Interacts with the regulatory GTPase-binding domain of N-WASP; inhibits activation of Arp2/3 complex by maintaining N-WASP in an inactive conformation. Also inhibits PIP2-induced actin polymerization (EC50 ∽ 4μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 426.15 |
| Purity | >99% |
| Molecular Weight (g/mol) | 426.15 |
| Chemical Name or Material | Wiskostatin |
Safety and Handling
| Recommended Storage | Store at +4°C |