accessibility menu, dialog, popup

UserName

H3CNCH3OHNBrBr

CAS RN 253449-04-6

IUPAC Name: 1-(3,6-dibromo-9H-carbazol-9-yl)-3-(dimethylamino)propan-2-ol
Synonyms: (S)-wiskostatin
Molecular Weight (g/mol): 426.15
Molecular Formula: C17H18Br2N2O
InChi Key: XUBJEDZHBUPBKL-UHFFFAOYNA-N
SMILES: CN(C)CC(O)CN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results