missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Wide Mouth Round Bottles with White Polypropylene Caps

Ideal for use with water based chemicals and some food products. Fisherbrand™ Wide Mouth Round Bottles with White Polypropylene Caps feature a wide opening making them suitable for solids.

 View more versions of this product

Catalog No. FB02912022A


Only null left

Description

Description

  • Ideal for general lab use with solids and powders
  • Provide an excellent moisture barrier, are translucent, and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and rough handling during shipping
  • Translucent bottles allow limited viewing of contents
Specifications

Chemical Identifiers

Natural
White
Wide
Polyethylene Foam
HDPE
Yes
Round Wide Mouth Bottle
Polypropylene

Specifications

250 mL
50 mm
179 mm
260
53-400
Wide
Round Wide Mouth Bottle
Polypropylene
67 mm
Natural
41 mm
HDPE
White
Yes
103 mm
Polyethylene Foam
53-400 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions