missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ Wide Mouth Oblong Plastic Bottle

Catalog No. 03084215
Change view
Click to view available options
Capacity (Metric):
120 mL
250 mL
Closure Included:
Yes
Quantity:
48 Bottles/Cs.
72 Bottles/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Included Quantity
03084215 250 mL Yes 48 Bottles/Cs.
01258106 120 mL - 72 Bottles/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03084215 Supplier DWK Life Sciences™ Supplier No. W218814

These wide mouth, oblong plastic bottles are made of natural color HDPE.

  • Available in a range of sizes and quantities to meet your needs

Specifications

Capacity (Metric) 250 mL
Color Natural
Material HDPE
Quantity 48 Bottles/Cs.
Closure Included Yes
Mouth Wide
Closure Type Screw Cap
Capacity (English) 8.5 oz.
Shape Oblong
Product Type Wide Mouth Bottle