missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Precisionware™ High-Density Polyethylene Wide-Mouth Bottles

Catalog No. 08647740
Change view
Click to view available options
32 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
08647740 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08647740 Supplier Bel-Art™ Supplier No. 106259018
Only null left

Ideal for packaging and storing of liquids and solids.

  • Rigid, leakproof design with easy-to-fill wide mouth
  • High-density polyethylene bottles with polypropylene closures
  • Feature recycling code molded into base, except S95194

Specifications

Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Translucent
Material Polypropylene
Packaging 75/Cs.
Quantity 32 oz.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Wide Mouth Bottle
Autoclavable Not Autoclavable
Show More Show Less