missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific Bench-Top Keeper™ Biohazard Container

Catalog No. 14375266
Change view
Click to view available options
Height (Metric):
18 cm
25 cm
Length (Metric):
10 cm
15 cm
Width (Metric):
10 cm
15 cm
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Length (Metric) Width (Metric) Size
14375266 25 cm 15 cm 15 cm Large
14375265 18 cm 10 cm 10 cm Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14375266 Supplier Heathrow Scientific Supplier No. BH2003
Edge

Economical point-of-use alternative to hard-sided sharps containers for disposal of contaminated Petri dishes, plastic assay vials, analyzer cups and other non-sharp waste

  • Made of sturdy paperboard with an internal plastic lining and webbed corners to contain moderate amounts of fluids
  • Self-locking tops keep contents safely enclosed
  • Small Keepers fold out to 4L x 4W x 7H in. (10 x 10 x 18 cm) and accommodate up to 1600 cc of material
  • Large Keepers fold out to 6L x 6W x 10H in. (15 x 15 x 25 cm) and accommodate up to 5000 cc of material
  • Autoclavable
  • Made in USA

Specifications

Capacity (Metric) 3 L
Capacity (English) 3000 cc
Dimensions (L x W x H) 15 x 15 x 25 cm (6 x 6 x 10 in.)
Disposable Single-use
Height (Metric) 25 cm
Length (Metric) 15 cm
Size Large
Width (Metric) 15 cm
Autoclavable Autoclavable
Includes Self-locking tops
Liner Material Plastic
For Use With (Application) Storing contaminated lab waste, disposal of contaminated petri dishes, plastic assay vials, analyzer cups and other non-sharp waste
Width (English) 6 in.
Length (English) 6 in.
Height (English) 10 in.
Material Paperboard
Product Type Benchtop Keeper
Show More Show Less