Learn More
Fisherbrand™ White Polypropylene Caps
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 1700 |
| Sterility | Non-sterile |
| Liner Material | Unlined |
| Neck Finish | 48-400 |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Product Type | Lined Cap |