Learn More
Fisherbrand™ White Polypropylene Caps
Fisherbrand™ White Polypropylene Caps include a clear writing surface for easy identification.
 Afficher plus de versions de ce produit
Numéro de catalogue. FB02912088A
Il en reste null
Description
Description
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Spécifications
Identifiants chimiques
White
Polypropylene
Spécifications
White
144
70-400
Polypropylene
Pulp/Vinyl
Lined Cap
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS