Learn More
Socorex Acura™ Manual Pipets 826 XS TwiXS pack from DWK Life Sciences™
Two adjustable volume pipettes available in six different packs
Supplier: Socorex W810313TXS
Includes: Shelf pipette holder, pipette tips samples, silicone grease, Q.C. certificates and operating instructions
Description
- Two adjustable volume pipettes
- Free shelf pipette holder included
- Volume combinations up to 1000μL
- Six different packs available
Chemical Identifiers
| CE, IVD 98/79 EEC | |
| 826 XS | |
| Ergonomic | |
| Yes |
| Autoclavable | |
| Acura | |
| Shelf pipette holder, pipette tip samples, Silicone Grease, Q.C. certificates and operating instructions | |
| 3 Years |
Specifications
| CE, IVD 98/79 EEC | |
| 826 XS | |
| Manual Pipettes | |
| Ergonomic | |
| Yes |
| Autoclavable | |
| Acura | |
| 2 to 20, 20 to 200 μL | |
| Shelf pipette holder, pipette tip samples, Silicone Grease, Q.C. certificates and operating instructions | |
| 3 Years |