missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Prescored Clear Standards Ampules
Glass containers designed to be hermetically sealed, preserving purity and extending content shelf life
$1,147.54
Spécifications
| Material | Borosilicate Glass |
|---|---|
| Product Type | Prescored Ampule |
| Color | Clear |
| Certifications/Compliance | ASTM E438 Type I, Class A |
| Numéro de catalogue | Numéro du manufacturier. | Capacity (Metric) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Capacity (Metric) | Prix | Quantity & Availability | |||||
|
01215E
|
DWK Life Sciences™
176759 |
7 mL |
caisse de 25 chaque for $1,147.54
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
A gold band on the stem indicates that the ampule is pre-scored to facilitate snapping.
- Constructed of USP Type I clear and amber low-extractable borosilicate glass
- Methods of sterilization: autoclave, dry heat and EtO
- Prevents cross contamination
Standards packaging, environmental and diagnostic standards, lyophilization and cryogenic storage
Spécifications
| Borosilicate Glass | |
| Prescored Ampule | |
| Clear | |
| ASTM E438 Type I, Class A |