missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Embouts de compte-gouttes contrôlés allongés à faible densité (15 mm)

Numéro de catalogue. 12141603
Modifier l'affichage
Click to view available options
Color:
Naturel
Orange
Material:
LDPE
PEBD
No. per Case:
100
1000 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color Material No. per Case
12141603 Orange LDPE 100
0344047 Naturel PEBD 1000 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 12141603 Fournisseur DWK Life Sciences Code fournisseur 242423

Pointe lance-gouttes contrôlée allongée de 15 mm en LDPE (polyéthylène de basse densité)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100

Spécifications

Color Orange
For Use With (Application) Controlled Liquid Dispensing, Dropwise Addition
Material LDPE
Length (Metric) 15 mm
No. per Case 100
Sterility Non-sterile
Disposable Reusable
Fit for Purpose Certified Quality
Product Type Dropper Tip
For Use With (Equipment) Wheaton (15-415) dropper caps and Wheaton dropping bottles
Autoclavable Not Autoclavable
Afficher plus de résultats Afficher moins de résultats