Learn More
DWK LIFE SCIENCES™ WHEATON™ Three-ball Snyder Column
Description
- Designed for concentration of trace amounts of analyte in organic solvent
- 24/40 Clear-Seal joint for secure, reliable connections
- Manufactured from borosilicate glass
- Three-ball design aids solvent reflux and analyte concentration efficiency
Specifications
Specifications
| For Use With (Equipment) | Kuderna-Danish Apparatus |
| Product Type | Three-ball Snyder Column |
| Includes | 24/40 clear seal joint |
| For Use With (Application) | Concentration of trace amounts of analyte in organic solvent |
| Certifications/Compliance | ASTM E 438 |