Learn More
DWK Life Sciences Wheaton™ Jars with Polypropylene Screw Cap
$262.43 - $645.68
Specifications
| Includes | Screw Cap |
|---|---|
| Closure Color | White |
| Mouth | Wide |
| Shape | Round |
| Product Type | Jar |
| Catalog Number | Mfr. No. | Capacity (Metric) | Autoclavable | Color | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Autoclavable | Color | Price | Quantity & Availability | |||
|
02912992
|
DWK Life Sciences
W209912 |
30 mL | Not Autoclavable | Clear |
Case of 72 Each for $344.86
|
|
|||
|
02912986
|
DWK Life Sciences
W209906 |
30 mL | Autoclavable (Bottle Only) | Natural |
Case of 72 Each for $330.75
|
|
|||
|
02912993
|
DWK Life Sciences
W209913 |
60 mL | Not Autoclavable | Clear |
Case of 48 Each for $310.29
|
|
|||
|
02912987
|
DWK Life Sciences
W209907 |
60 mL | Not Autoclavable | Natural |
Case of 48 Each for $303.04
|
|
|||
|
02912981
|
DWK Life Sciences
W209901 |
60 mL | Autoclavable (Bottle Only) | White |
Case of 48 Each for $277.86
|
|
|||
|
02912988
|
DWK Life Sciences
W209908 |
125 mL | Not Autoclavable | Natural |
Case of 36 Each for $262.43
|
|
|||
|
02912994
|
DWK Life Sciences
W209914 |
125 mL | Not Autoclavable | Clear |
Case of 36 Each for $286.82
|
|
|||
|
02912995
|
DWK Life Sciences
W209915 |
250 mL | Not Autoclavable | Clear |
Case of 36 Each for $436.29
|
|
|||
|
02912989
|
DWK Life Sciences
W209909 |
250 mL | Not Autoclavable | Natural |
Case of 36 Each for $455.96
|
|
|||
|
02912983
|
DWK Life Sciences
W209903 |
250 mL | Autoclavable (Bottle Only) | White |
Case of 36 Each for $430.04
|
|
|||
|
02912990
|
DWK Life Sciences
W209910 |
500 mL | Autoclavable (Bottle Only) | Natural |
Case of 24 Each for $373.29
|
|
|||
|
02912996
|
DWK Life Sciences
W209916 |
500 mL | Not Autoclavable | Clear |
Case of 24 Each for $381.39
|
|
|||
|
02912984
|
DWK Life Sciences
W209904 |
500 mL | Autoclavable (Bottle Only) | White |
Case of 24 Each for $343.68
|
|
|||
|
02912991
|
DWK Life Sciences
W209911 |
1000 mL | Not Autoclavable | Natural |
Case of 24 Each for $624.32
|
|
|||
|
02912997
|
DWK Life Sciences
W209917 |
1000 mL | Not Autoclavable | Clear |
Case of 24 Each for $634.43
|
|
|||
|
02912985
|
DWK Life Sciences
W209905 |
1000 mL | Autoclavable (Bottle Only) | White |
Case of 24 Each for $645.68
|
|
|||
Description
- Good resistance to inorganic chemicals
- Pre-attached white polypropylene screw cap with foamed polyethylene liner
Specifications
| Screw Cap | |
| Wide | |
| Jar | |
| Screw Cap |
| White | |
| Round | |
| Polypropylene |