missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ WHEATON™ Headspace Magnetic Seal

Catalog No. 03410727
Change view
Click to view available options
Color:
Gold
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03410727 Gold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03410727 Supplier DWK LIFE SCIENCES™ Supplier No. W224223

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

WHEATON™ Headspace Magnetic Seal is designed for use with crimp top headspace vials. Featuring a 20mm aluminum construction with PTFE/Silicone septa, this gold-colored magnetic seal provides a reliable, secure closure for headspace analysis applications.

  • WheatonTM Headspace Magnetic Seal for crimp top headspace vials
  • 20mm size for compatibility with standard headspace vials
  • Aluminum construction with gold color finish
  • PTFE/Silicone septa for reliable sealing performance
  • Magnetic design for secure closure during headspace analysis

Specifications

Color (Seal) Gold
Color Gold
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum PTFE/Silicone
Closure Type Headspace Magnetic Seal
Quantity 100/Cs.
For Use With (Equipment) Crimp Top Headspace Vials
Autoclavable Not Autoclavable
Product Type Magnetic Crimp Cap
Show More Show Less