missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ FTA™ Nucleic Acid Collection Storage box

Catalog No. 02912752
Change view
Click to view available options
Dimensions (L x W x H):
18 x 7 x 4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
02912752 18 x 7 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02912752 Supplier DWK Life Sciences™ Supplier No. W651612

For nucleic acid collection

  • Accommodates two rows of 3 in. FTA card pouches or one row of 6 in. FTA Card Pouches
  • Removable divider
  • Water repellent material

Specifications

Type Nucleic Acid Collection Storage Box
Dimensions (L x W x H) 18 x 7 x 4 in.