Learn More
DWK Life Sciences™ Wheaton™ Cryule™ Gold-Band™ Cryogenic Ampules
For preservation of biological materials with liquid nitrogen
Supplier:  DWK Life Sciences™ 651486
 Afficher plus de versions de ce produit
Numéro de catalogue. 06401D
Il en reste null
Avertissement:
Cryogenic vials are intended for placement only in the vapor phase of liquid nitrogen, and should not be used for storage in the liquid phase of liquid nitrogen. Immersion of the vials in the liquid phase could result in penetration of the liquefied gas into the vial, resulting in rapid vaporization of the liquid upon removal and possible violent explosion or leakage from the vial/closure perimeter.
To prevent cryogenic vials from exploding, never overfill liquid nitrogen storage units. Always examine vials before use to ensure no visible defects around the closure rims. Always use full faceshields, heavy safety gloves and laboratory protective apparel when removing vials from cryogenic storage. Always permit vials to warm slowly in a biological safety cabinet. Never reuse cryogenic vials.
Description
Description
  • Low-extractable borosilicate glass
  • Gold-Band feature indicates prescoring
  • Tips of prescored ampules snap off easily without filing
  • Designed for low-temperature storage and rapid thawing
Spécifications
Identifiants chimiques
Borosilicate Glass
Clear
ASTM E438 Type I
Spécifications
2 mL
Prescored Cryogenic Ampule
1.15 cm
7 cm
0.5 cm
ASTM E438 Type I
Borosilicate Glass
Clear
0.45 in.
2.75 in.
0.19 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires