Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Supplier:  DWK Life Sciences W991009CP
 Afficher plus de versions de ce produit
Numéro de catalogue. 06451229A
Il en reste null
Description
Description
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Spécifications
Identifiants chimiques
Sterile
Vial, Stopper, and Seal
Wheaton CompletePAK
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Spécifications
Red
Butyl, Glass
5 mL
120/Cs.
Ultra Pure Serum Stopper
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Clear
Sterile
Vial, Stopper, and Seal
Wheaton CompletePAK
20 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS