missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

$2,783.47 - $6,711.65

Specifications

Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
View More Specs

Products 18
Catalog Number Mfr. No. Closure Color Closure Type Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Closure Color Closure Type Volume (Metric) Price Quantity & Availability  
15112152A
View Documents Promotion Details
DWK Life Sciences
W991002CPA
Blue OmniFlex 3G Igloo Lyo Stopper 2 mL
Case of 278 Each for $6,711.65
Only null left
 
15112155A
View Documents Promotion Details
DWK Life Sciences
W991005CPA
Red OmniFlex 3G Igloo Lyo Stopper 2 mL
Case of 278 Each for $6,711.65
Only null left
 
15112151A
View Documents Promotion Details
DWK Life Sciences
W991001CPA
Blue OmniFlex 3G Serum Stopper 2 mL
Case of 278 Each for $6,404.56
Only null left
 
15112154A
View Documents Promotion Details
DWK Life Sciences
W991004CPA
Red OmniFlex 3G Serum Stopper 2 mL
Case of 278 Each for $6,404.56
Only null left
 
15112150A
View Documents Promotion Details
DWK Life Sciences
W991000CPA
Blue Ultra Pure Serum Stopper 2 mL
Case of 278 Each for $5,848.88
Only null left
 
15112153A
View Documents Promotion Details
DWK Life Sciences
W991003CPA
Red Ultra Pure Serum Stopper 2 mL
Case of 278 Each for $5,848.88
Only null left
 
15112158A
View Documents Promotion Details
DWK Life Sciences
W991008CPA
Blue OmniFlex 3G Igloo Lyo Stopper 5 mL
Case of 120 Each for $3,114.82
Only null left
 
15112161A
View Documents Promotion Details
DWK Life Sciences
W991011CPA
Red OmniFlex 3G Igloo Lyo Stopper 5 mL
Case of 120 Each for $3,114.82
Only null left
 
15112157A
View Documents Promotion Details
DWK Life Sciences
W991007CPA
Blue OmniFlex 3G Serum Stopper 5 mL
Case of 120 Each for $3,095.94
Only null left
 
15112160A
View Documents Promotion Details
DWK Life Sciences
W991010CPA
Red OmniFlex 3G Serum Stopper 5 mL
Case of 120 Each for $3,035.35
Only null left
 
15112156A
View Documents Promotion Details
DWK Life Sciences
W991006CPA
Blue Ultra Pure Serum Stopper 5 mL
Case of 120 Each for $2,783.47
Only null left
 
15112159A
View Documents Promotion Details
DWK Life Sciences
W991009CPA
Red Ultra Pure Serum Stopper 5 mL
Case of 120 Each for $2,783.47
Only null left
 
15112164A
View Documents Promotion Details
DWK Life Sciences
W991014CPA
Blue OmniFlex 3G Igloo Lyo Stopper 10 mL
Case of 104 Each for $3,633.76
Only null left
 
15112167A
View Documents Promotion Details
DWK Life Sciences
W991017CPA
Red OmniFlex 3G Igloo Lyo Stopper 10 mL
Case of 104 Each for $3,562.59
Only null left
 
15112163A
View Documents Promotion Details
DWK Life Sciences
W991013CPA
Blue OmniFlex 3G Serum Stopper 10 mL
Case of 104 Each for $3,544.76
Only null left
 
15112166A
View Documents Promotion Details
DWK Life Sciences
W991016CPA
Red OmniFlex 3G Serum Stopper 10 mL
Case of 104 Each for $3,475.26
Only null left
 
15112162A
View Documents Promotion Details
DWK Life Sciences
W991012CPA
Blue Ultra Pure Serum Stopper 10 mL
Case of 104 Each for $3,262.85
Only null left
 
15112165A
View Documents Promotion Details
DWK Life Sciences
W991015CPA
Red Ultra Pure Serum Stopper 10 mL
Case of 104 Each for $3,262.94
Only null left
 
Description

Description

  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions
Specifications

Specifications

Amber
No
Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Vial, Stopper, and Seal
Wheaton CompletePAK
Borosilicate Glass
Sterile
No
Crimp top
No
Videos
SDS
Documents

Documents