missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Supplier: DWK Life Sciences W991006CPA
Description
- Products supplied ready-to-use
- Reduces lead times
- Focuses your efforts on core competencies
- Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
- Frees up valuable clean room space
- Limits number of suppliers required to provide complete packaging product
- Amber for photosensitive contents
- Validated processes
- Saves valuable time and money while maintaining a high quality product and streamlining process
- One source for total packaging solutions
Chemical Identifiers
| Amber | |
| No | |
| Light Sensitive Parenteral Preparations and Ready-to-Use Applications | |
| Vial, Stopper, and Seal | |
| Wheaton CompletePAK |
| Borosilicate Glass | |
| Sterile | |
| No | |
| Crimp top | |
| No |
Specifications
| Blue | |
| Borosilicate Glass | |
| Sterile | |
| Light Sensitive Parenteral Preparations and Ready-to-Use Applications | |
| Vial, Stopper, and Seal | |
| Crimp top | |
| Ultra Pure Serum Stopper | |
| No |
| Amber | |
| No | |
| 5 mL | |
| No | |
| 120/Cs. | |
| Wheaton CompletePAK | |
| 20 mm | |
| 40 mm |