missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits

Catalog No. 15112152A
Change view
Click to view available options
Closure Color:
Blue
Red
Closure Type:
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure Serum Stopper
Volume (Metric):
2 mL
5 mL
10 mL
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Closure Type Volume (Metric)
15112152A Blue OmniFlex 3G Igloo Lyo Stopper 2 mL
15112155A Red OmniFlex 3G Igloo Lyo Stopper 2 mL
15112151A Blue OmniFlex 3G Serum Stopper 2 mL
15112154A Red OmniFlex 3G Serum Stopper 2 mL
15112150A Blue Ultra Pure Serum Stopper 2 mL
15112153A Red Ultra Pure Serum Stopper 2 mL
15112158A Blue OmniFlex 3G Igloo Lyo Stopper 5 mL
15112161A Red OmniFlex 3G Igloo Lyo Stopper 5 mL
15112157A Blue OmniFlex 3G Serum Stopper 5 mL
15112160A Red OmniFlex 3G Serum Stopper 5 mL
15112156A Blue Ultra Pure Serum Stopper 5 mL
15112159A Red Ultra Pure Serum Stopper 5 mL
15112164A Blue OmniFlex 3G Igloo Lyo Stopper 10 mL
15112167A Red OmniFlex 3G Igloo Lyo Stopper 10 mL
15112163A Blue OmniFlex 3G Serum Stopper 10 mL
15112166A Red OmniFlex 3G Serum Stopper 10 mL
15112162A Blue Ultra Pure Serum Stopper 10 mL
15112165A Red Ultra Pure Serum Stopper 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 15112152A Supplier DWK Life Sciences Supplier No. W991002CPA

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions

Specifications

Closure Color Blue
Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Volume (Metric) 2 mL
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Graduated No
Includes Vial, Stopper, and Seal
No. per Case 278/Cs.
Style Crimp top
Product Line Wheaton CompletePAK
Closure Type OmniFlex 3G Igloo Lyo Stopper
Closure Size 13 mm
Marking Spot No
Height (Metric) 32 mm
Show More Show Less