Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Supplier:  DWK Life Sciences W991008CPA
 View more versions of this product
Catalog No. 15112158A
Only null left
Description
Description
  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions
Specifications
Chemical Identifiers
Amber
No
Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Vial, Stopper, and Seal
Wheaton CompletePAK
Borosilicate Glass
Sterile
No
Crimp top
No
Specifications
Blue
Borosilicate Glass
Sterile
Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Vial, Stopper, and Seal
Crimp top
OmniFlex 3G Igloo Lyo Stopper
No
Amber
No
5 mL
No
120/Cs.
Wheaton CompletePAK
13 mm
40 mm
Product Suggestions
Product Suggestions