Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars without Caps
Ideal for soil sampling and environmental applications
$63.14 - $426.00
Specifications
Color Clear
Material Soda Lime Glass
Closure Type Screw Cap
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
02912190
View Documents Promotion Details
DWK Life Sciences
W216919
60 mL 53-400 mm
Case of 144 Each for $426.00
Only null left
02912191
View Documents Promotion Details
DWK Life Sciences
W216920
125 mL 58-400 mm
Case of 24 Each for $76.29
Only null left
02912192
View Documents Promotion Details
DWK Life Sciences
W216921
250 mL 70-400 mm
Case of 24 Each for $100.75
Only null left
02912193
View Documents Promotion Details
DWK Life Sciences
W216922
500 mL 89-400 mm
Case of 12 Each for $63.14
Only null left
02912194
View Documents Promotion Details
DWK Life Sciences
W216923
1000 mL 89-400 mm
Case of 12 Each for $98.39
Only null left
Description
Description
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
  • Wide mouths provide easy access to contents when using spoons or spatulas
  • Chemical-resistant caps conform to EPA and FDA regulations
  • Available in partitioned Convenience or Valu-Bulk™ cartons
  • Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper
  • Evident labels to protect sample integrity in transit or long-term storage
Specifications
Specifications
Clear
Soda Lime Glass
Screw Cap