missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Straight-Sided Jars without Caps

Catalog No. 02912194
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
53-400 mm
58-400 mm
70-400 mm
89-400 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
02912194 1000 mL 89-400 mm
02912190 60 mL 53-400 mm
02912191 125 mL 58-400 mm
02912192 250 mL 70-400 mm
02912193 500 mL 89-400 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02912194 Supplier DWK Life Sciences Supplier No. W216923

Ideal for soil sampling and environmental applications

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Wide mouths provide easy access to contents when using spoons or spatulas
  • Chemical-resistant caps conform to EPA and FDA regulations
  • Available in partitioned Convenience or Valu-Bulk™ cartons
  • Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper
  • Evident labels to protect sample integrity in transit or long-term storage

Specifications

Capacity (Metric) 1000 mL
Color Clear
Material Soda Lime Glass
Height (Metric) 170 mm
Height (English) 6.6 in.
Closure Size 89-400 mm
Diameter (Metric) 95 mm
Closure Type Screw Cap
Capacity (English) 32 oz.