missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Straight-Sided Jars without Caps

Ideal for soil sampling and environmental applications

Supplier:  DWK Life Sciences W216921

 View more versions of this product

Catalog No. 02912192


Only null left

Description

Description

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Wide mouths provide easy access to contents when using spoons or spatulas
  • Chemical-resistant caps conform to EPA and FDA regulations
  • Available in partitioned Convenience or Valu-Bulk™ cartons
  • Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper
  • Evident labels to protect sample integrity in transit or long-term storage

Specifications

Chemical Identifiers

Clear
Soda Lime Glass
Screw Cap

Specifications

250 mL
Clear
88 mm
70-400 mm
Screw Cap
8 oz.
Soda Lime Glass
3.4 in.
73 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions