Learn More
DWK Life Sciences WHEATON™ Borosilicate Glass Coliwasa Sampler Replacement PTFE Seals
Catalog No. 1368316
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
1368316 PTFE
1 options
Catalog No. 1368316 Supplier DWK Life Sciences Supplier No. 885232
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
WHEATON™ Borosilicate Glass Coliwasa Sampler Replacement PTFE Seals provide a positive PTFE seal for collecting composite liquid samples from top to bottom in drums, tanks, and reservoirs. Sold 10 per case.
  • Replacement PTFE seals for Wheaton borosilicate glass Coliwasa samplers
  • Positive PTFE seal design ensures reliable, leak-resistant closure
  • Compatible with Coliwasa samplers used to collect composite liquid samples from drums, tanks, and reservoirs
  • Sold 10 per case
Specifications
Type Seal, Replacement
Material PTFE
Length (Metric) 106.5 cm
Length (English) 42 in.
For Use With (Application) Used to collect composite liquid samples from top to bottom in drums, tanks and reservoirs
For Use With (Equipment) For Borosilicate Glass coliwasa