Learn More
DWK LIFE SCIENCES™ WHEATON™ Additional Decks for Modular R2P Roller Culture Apparatus
Catalog No. 02912629
Change view
Click to view available options
Product Type:
Deck Kit
Factory Installed
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
02912629 Deck Kit
02912628 Factory Installed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02912629 Supplier DWK LIFE SCIENCES™ Supplier No. W348887
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
WHEATON™ Additional Decks for Modular R2P Roller Culture Apparatus allow you to scale up from research to production quantities without having to change equipment or revalidate protocols. Each 5-position deck kit expands roller bottle capacity with modular spacing.
  • 5-position add-on deck kit for Wheaton Modular R2P Roller Culture Apparatus
  • Modular spacing of 7.106 in. deck-to-deck
  • Allows scale-up from research to production quantities without changing equipment or revalidating protocols
  • Maximum capacity of 9 decks per unit
  • Available factory-installed (must order with unit) or as a kit for customer installation
Specifications
Product Type Deck Kit
For Use With (Application) For customer installation
For Use With (Equipment) Modular R2P roller culture apparatus